- Chiral amorphous metal-organic polyhedra used as the stationary phase for high-resolution gas chromatography separations.
Chiral amorphous metal-organic polyhedra used as the stationary phase for high-resolution gas chromatography separations.
Chirality (2020-07-06)
Bo Tang, Chenyu Sun, Wei Wang, Lina Geng, Liquan Sun, Aiqin Luo
PMID32623797
ABSTRACT
Herein, we describe a new chiral amorphous metal-organic polyhedra used as the stationary phase for high-resolution gas chromatography (GC). The chiral stationary phase was coated onto a capillary column via a dynamic coating process and investigated for a variety of compounds. The experimental results showed that the chiral stationary phase exhibits good selectivity for linear alkanes, linear alcohols, polycyclic aromatic hydrocarbons, isomers, and chiral compounds. In addition, the column has the advantages of high column efficiency and short analysis time. The present work indicated that amorphous metal-organic polyhedra have great potential for application as a new type of stationary phase for GC.