Merck
  • 2,7-Diphenyl[1]benzothieno[3,2-b]benzothiophene, a new organic semiconductor for air-stable organic field-effect transistors with mobilities up to 2.0 cm2 V(-1) s(-1).
2,7-Diphenyl[1]benzothieno[3,2-b]benzothiophene, a new organic semiconductor for air-stable organic field-effect transistors with mobilities up to 2.0 cm2 V(-1) s(-1).
Journal of the American Chemical Society (2006-09-28)
Kazuo Takimiya, Hideaki Ebata, Katsuhiro Sakamoto, Takafumi Izawa, Tetsuo Otsubo, Yoshihito Kunugi
ABSTRACT
Vapor-deposited thin films of a newly developed sulfur-containing heteroarene, 2,7-diphenyl[1]benzothieno[3,2-b][1]benzothiophene (DPh-BTBT), were used as an active layer of OFETs, which showed excellent FET characteristics in ambient conditions with mobilities of approximately 2.0 cm2 V-1 s-1 and Ion/Ioff of 107.