Merck
  • Characterization of an Amphetamine Interference from Gabapentin in an LC-HRMS Method.
Characterization of an Amphetamine Interference from Gabapentin in an LC-HRMS Method.
Journal of analytical toxicology (2019-07-03)
Ana Celia Muñoz-Muñoz, Teresa Pekol, Dana Schubring, Robin Hyland, Charlene Johnson, Lawrence Andrade
ABSTRACT
An amphetamine interference was observed during the development of an liquid chromatography-high-resolution mass spectrometry (LC-HRMS) multi-class confirmation method for the determination of 47 drugs and metabolites in urine. The interference passed all qualitative criteria for amphetamine leading to potential false-positive results. Upon investigation, it was found that the amphetamine interference was correlated with the presence of high levels of gabapentin. Gabapentin is routinely detected in patient urine specimens at levels in excess of 1 mg/mL as it is widely prescribed at high doses and does not undergo significant metabolism. The source of the interference was identified as a gabapentin in-source fragment isomeric with protonated amphetamine. Here we describe the characterization of this interference and how its effect was mitigated in the LC-HRMS method.