Merck
  • [Relativity among starch quantity, polysaccharides content and total alkaloid content of Dendrobium loddigesii].
[Relativity among starch quantity, polysaccharides content and total alkaloid content of Dendrobium loddigesii].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-03-08)
Hua Zhu, Jianbei Teng, Yi Cai, Jie Liang, Yilin Zhu, Tao Wei
ABSTRACT
To find out the relativity among starch quantity, polysaccharides content and total alkaloid content of Dendrobium loddigesii. Microscopy-counting process was applied to starch quantity statistics, sulfuric acid-anthrone colorimetry was used to assay polysaccharides content and bromocresol green colorimetry was used to assay alkaloid content. Pearson product moment correlation analysis, Kendall's rank correlation analysis and Spearman's concordance coefficient analysis were applied to study their relativity. Extremely significant positive correlation was found between starch quantity and polysaccharides content, and significant negative correlation between alkaloid content and starch quantity was discovered, as well was between alkaloid content and polysaccharides content.
Sigma-Aldrich
Bromocresol Green sodium salt, ACS reagent