Merck
  • Semi-mechanistic modelling of the tumour growth inhibitory effects of LY2157299, a new type I receptor TGF-beta kinase antagonist, in mice.
Semi-mechanistic modelling of the tumour growth inhibitory effects of LY2157299, a new type I receptor TGF-beta kinase antagonist, in mice.
European journal of cancer (Oxford, England : 1990) (2007-11-28)
Lorea Bueno, Dinesh P de Alwis, Celine Pitou, Jonathan Yingling, Michael Lahn, Sophie Glatt, Iñaki F Trocóniz
ABSTRACT
Human xenografts Calu6 (non-small cell lung cancer) and MX1 (breast cancer) were implanted subcutaneously in nude mice and LY2157299, a new type I receptor TGF-beta kinase antagonist, was administered orally. Plasma levels of LY2157299, percentage of phosphorylated Smad2,3 (pSmad) in tumour, and tumour size were used to establish a semi-mechanistic pharmacokinetic/pharmacodynamic model. An indirect response model was used to relate plasma concentrations with pSmad. The model predicts complete inhibition of pSmad and rapid turnover rates [t(1/2) (min)=18.6 (Calu6) and 32.0 (MX1)]. Tumour growth inhibition was linked to pSmad using two signal transduction compartments characterised by a mean signal propagation time with estimated values of 6.17 and 28.7 days for Calu6 and MX1, respectively. The model provides a tool to generate experimental hypothesis to gain insights into the mechanisms of signal transduction associated to the TGF-beta membrane receptor type I.
Sigma-Aldrich
Galunisertib, ≥98% (HPLC)