Merck
  • Reactive Insertion of PEDOT-PSS in SWCNT@Silica Composites and its Electrochemical Performance.
Reactive Insertion of PEDOT-PSS in SWCNT@Silica Composites and its Electrochemical Performance.
Materials (Basel, Switzerland) (2020-03-12)
Halima Djelad, Abdelghani Benyoucef, Emilia Morallón, Francisco Montilla
ABSTRACT
Hybrid silica-modified materials were synthesized on glassy carbon (GC) electrodes by electroassisted deposition of sol-gel precursors. Single-wall carbon nanotubes (SWCNTs) were dispersed in a silica matrix (SWCNT@SiO2) to enhance the electrochemical performance of an inorganic matrix. The electrochemical behavior of the composite electrodes was tested against the ferrocene redox probe. The SWCNT@SiO2 presents an improvement in the electrochemical performance towards ferrocene. The heterogeneous rate constant of the SWCNT@SiO2 can be enhanced by the insertion of poly(3,4-Ethylendioxythiophene)-poly(sodium 4-styrenesulfonate) PEDOT-PSS within the silica matrix, and this composite was synthesized successfully by reactive electrochemical polymerization of the precursor EDOT in aqueous solution. The SWCNT@SiO2-PEDOT-PSS composite electrodes showed a heterogeneous rate constant more than three times higher than the electrode without conducting polymer. Similarly, the electroactive area was also enhanced to more than twice the area of SWCNT@SiO2-modified electrodes. The morphology of the sample films was analyzed by scanning electron microscopy (SEM).
Sigma-Aldrich
3,4-Ethylenedioxythiophene, 97%
Sigma-Aldrich
Sodium 4-vinylbenzenesulfonate, technical, ≥90% (T)