Merck
  • Metabolic engineering of Escherichia coli to produce succinate from woody hydrolysate under anaerobic conditions.
Metabolic engineering of Escherichia coli to produce succinate from woody hydrolysate under anaerobic conditions.
Journal of industrial microbiology & biotechnology (2020-01-29)
Fayin Zhu, Chengqiang Wang, Ka-Yiu San, George N Bennett
ABSTRACT
It is of great economic interest to produce succinate from low-grade carbon sources, e.g., lignocellulosic biomass hydrolysate, which mainly contains glucose and xylose. Inactivation of the glucose uptake system PtsG was evaluated for succinate production from xylose-rich feedstocks. Strains with integration of succinate production modules into the chromosome of Escherichia coli were then constructed. These strains have better succinate production performance from xylose-rich feedstocks than strain FZ560 harboring pHL413KF1. Glucose utilization was enhanced in FZ661T by manipulation of the gal operon to allow efficient use of the high-concentration glucose in woody biomass hydrolysate. Up to 906.7 mM (107.0 g/L) succinate was produced from mixed sugars in fed-batch fermentation and more than 461.7 mM (54.5 g/L) succinate was produced from woody hydrolysate in a batch fermentation. In this study, FZ661T was able to produce succinate from woody hydrolysate in minimal medium efficiently, making it attractive for industrial applications in succinate production.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
L-(−)-Xylose, mixture of anomers, ≥99%
Pricing and availability is not currently available.