Merck
  • Anatomical and single-cell transcriptional profiling of the murine habenular complex.
Anatomical and single-cell transcriptional profiling of the murine habenular complex.
eLife (2020-02-12)
Michael L Wallace, Kee Wui Huang, Daniel Hochbaum, Minsuk Hyun, Gianna Radeljic, Bernardo L Sabatini
ABSTRACT
The lateral habenula (LHb) is an epithalamic brain structure critical for processing and adapting to negative action outcomes. However, despite the importance of LHb to behavior and the clear anatomical and molecular diversity of LHb neurons, the neuron types of the habenula remain unknown. Here, we use high-throughput single-cell transcriptional profiling, monosynaptic retrograde tracing, and multiplexed FISH to characterize the cells of the mouse habenula. We find five subtypes of neurons in the medial habenula (MHb) that are organized into anatomical subregions. In the LHb, we describe four neuronal subtypes and show that they differentially target dopaminergic and GABAergic cells in the ventral tegmental area (VTA). These data provide a valuable resource for future study of habenular function and dysfunction and demonstrate neuronal subtype specificity in the LHb-VTA circuit.
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation