Merck
  • Observation of the keto tautomer of D-fructose in D(2)O using (1)H NMR spectroscopy.
Observation of the keto tautomer of D-fructose in D(2)O using (1)H NMR spectroscopy.
Carbohydrate research (2011-12-02)
Thomas Barclay, Milena Ginic-Markovic, Martin R Johnston, Peter Cooper, Nikolai Petrovsky
ABSTRACT
D-Fructose was analysed by NMR spectroscopy and previously unidentified (1)H NMR resonances were assigned to the keto and α-pyranose tautomers. The full assignment of shifts for the various fructose tautomers enabled the use of (1)H NMR spectroscopy in studies of the mutarotation (5-25°C) and tautomeric composition at equilibrium (5-50°C). The mutarotation of β-pyranose to furanose tautomers in D(2)O at a concentration of 0.18 M was found to have an activation energy of 62.6 kJmol(-1). At tautomeric equilibrium (20°C in D(2)O) the distribution of the β-pyranose, β-furanose, α-furanose, α-pyranose and the keto tautomers was found to be 68.23%, 22.35%, 6.24%, 2.67% and 0.50%, respectively. This tautomeric composition was not significantly affected by varying concentrations between 0.089 and 0.36 M or acidification to pH 3. Upon equilibrating at 6 temperatures between 5 and 50°C there was a linear relationship between the change in concentration and temperature for all forms.
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D