Merck
  • Pulchinenosides from Pulsatilla Chinensis Increase P-Glycoprotein Activity and Induce P-Glycoprotein Expression.
Pulchinenosides from Pulsatilla Chinensis Increase P-Glycoprotein Activity and Induce P-Glycoprotein Expression.
Evidence-based complementary and alternative medicine : eCAM (2020-03-10)
Yali Liu, Ling Zhang, Shaofeng Wei, Jinyang Cai, Zhenzhong Zang, Meng Wang, Dan Su, Phillip M Gerk
ABSTRACT
Five pulchinenosides (pulchinenoside B3, pulchinenoside BD, pulchinenoside B7, pulchinenoside B10, and pulchinenoside B11) isolated from Pulsatilla chinensis (Bge) Regel saponins extract exhibited strong antitumor activities but poor gastrointestinal absorption properties. The enteric induction of P-glycoprotein (P-gp) is understood to restrict the oral bioavailability of some pharmaceutical compounds and lead to adverse drug reactions. Therefore, the present investigation was intended to delineate the impacts of pulchinenosides on cellular P-gp function and expression using Sf9 membrane vesicles and LS180 cells as a surrogate of human intestinal epithelial cells. Preliminary cytotoxic studies showed that 10 μM was an acceptable concentration for cytotoxicity and antiproliferation studies for all pulchinenosides using the alamarBlue assay. The cell cycle of LS180 cells detected by flow cytometry was not significantly influenced after 48 hours of coincubation with 10 μM of pulchinenosides. In the presence of pulchinenosides, the ATP-dependent transport of N-methyl-quinidine mediated by P-glycoprotein was stimulated significantly. The upregulation of P-glycoprotein and mRNA levels was found by Western blot and real-time PCR analysis in LS180 cells. Parallel changes indicate that all pulchinenosides are exposed to pulchinenosides-mediated transcriptional regulation. In conclusion, pulchinenosides could induce P-glycoprotein expression and directly increase its functional activity.
Sigma-Aldrich
Tetrabromophenol Blue, Dye content 75 %