Merck
  • Temporal dynamics of Arc/Arg3.1 expression in the dorsal striatum during acquisition and consolidation of a motor skill in mice.
Temporal dynamics of Arc/Arg3.1 expression in the dorsal striatum during acquisition and consolidation of a motor skill in mice.
Neurobiology of learning and memory (2020-01-07)
Wan-Kun Gong, Jian Ni, Lan-Fang Yu, Lu Wang, Zhi-Li Huang
ABSTRACT
Region- and pathway-specific plasticity within striatal circuits is critically involved in the acquisition and long-term retention of a new motor skill as it becomes automatized. However, the molecular substrates contributing to this plasticity remain unclear. Here, we examined the expression of the activity-regulated cytoskeleton-associated protein (Arc) in the associative or dorsomedial striatum (DMS) and the sensorimotor or dorsolateral striatum (DLS), as well as in striatonigral and striatopallidal neurons, during different skill learning phases in the accelerating rotarod task. We found that Arc was mainly expressed in the DMS during early motor learning and progressively increased in the DLS during gradual motor skill consolidation. Moreover, Arc was preferentially expressed in striatopallidal neurons early in training and gradually increased in striatonigral neurons later in training. These data demonstrate that in the dorsal striatum, the expression of Arc exhibits a region- and cell-specific transfer during the learning of a motor skill, suggesting a link between striatal Arc expression and motor skill learning in mice.
Sigma-Aldrich
ExtrAvidin®−FITC, buffered aqueous solution