Merck
  • TOPK promotes epithelial-mesenchymal transition and invasion of breast cancer cells through upregulation of TBX3 in TGF-β1/Smad signaling.
TOPK promotes epithelial-mesenchymal transition and invasion of breast cancer cells through upregulation of TBX3 in TGF-β1/Smad signaling.
Biochemical and biophysical research communications (2019-11-24)
Young-Ju Lee, Jung-Hwan Park, Sang-Muk Oh
ABSTRACT
TOPK has been suggested to contribute to invasion of lung, prostate, gastric, pancreatic or breast cancer cells. However, how TOPK mediates TGF-β1/Smad signaling leading to epithelial-mesenchymal transition (EMT) and invasion of breast cancer cells remains unknown. Here we report that TOPK upregulates T-box transcription factor TBX3 to enhance TGF-β1-induced EMT and invasion of MDA-MB-231 breast cancer cells. Expression of endogenous TOPK was promoted by TGF-β1 treatment of MDA-MB-231 cells time-dependently. In addition, knockdown of TOPK attenuated TGF-β1-induced phosphorylation or transcriptional activity of Smad3. Meanwhile, levels of both mRNA and protein of TBX3 induced by TGF-β1 were abolished by TOPK depletion. Also, knockdown of TBX3 inhibited TGF-β1 induction of EMT-related genes Snail, Slug or Fibronectin. Furthermore, ablation of TOPK or TBX3 suppressed TGF-β1-induced MDA-MB-231 cell invasion. Collectively, we conclude that TOPK positively regulates TBX3 in TGF-β1/Smad signaling pathway, thereby enhancing EMT and invasion of breast cancer cells, implying a mechanistic role of TOPK in TGF-β1/Smad signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting human PBK
Sigma-Aldrich
MISSION® esiRNA, targeting human TBX3