Merck
  • Icariin Ameliorates Amyloid Pathologies by Maintaining Homeostasis of Autophagic Systems in Aβ1-42-Injected Rats.
Icariin Ameliorates Amyloid Pathologies by Maintaining Homeostasis of Autophagic Systems in Aβ1-42-Injected Rats.
Neurochemical research (2019-10-16)
Xia Jiang, Lin-Lin Chen, Zhou Lan, Fan Xiong, Xiang Xu, Yang-Yang Yin, Ping Li, Ping Wang
ABSTRACT
Macroautophagy, a sole pathway for dysfunctional organelles or aggregated proteins turnover, has been implicated in the early development of Alzheimer's disease (AD). Previous studies have found that reversal of autophagy dysfunction in APP transgenic mice ameliorates amyloid pathologies. Icariin (ICA), the main component from traditional Chinese herb Epimedium brevicornu Maxim., can reduce accumulations of amyloid-β (Aβ) peptide in vivo and in vitro, but the mechanism remains unclear. Here, we explored the effects of ICA on autophagy-lysosomal pathway in intracerebroventricular (icv) injection of human Aβ1-42 peptide rats. We demonstrated that feeding the rats with ICA (30 mg/kg, 60 mg/kg and 90 mg/kg rat, per os) for 4 weeks rescued the Aβ1-42-induced spatial memory impairments, reduced endogenous rat Aβ42 tested by ELISA and decreased Aβ accumulation using 6E10 antibody. Furthermore, Aβ1-42 induced strong autophagy response, however ICA decreased the levels of microtubule-associated protein 1 light chain 3 (LC3) II/LC3I, Beclin1, Cathepsin D (Cat D) and brain lysosomal Cathepsin D activity. We also observed that ICA enhanced the phosphorylation of protein kinase B (PKB/AKT) and p70 ribosomal protein S6 kinase (p70S6K). In addition, ICA arrested Aβ1-42-induced cells loss, mitochondrias damage, nuclear membranes unclear and abundant nucleas chromatin agglutinates in hippocampus, lessened the expression of Cleaved-caspase-3, brain oxidative stress, astroglial activation. These findings suggest that ICA can ameliorate amyloid pathologies with improving autophagy-lysosome function and Chinese materia medica may be potential for AD treatment.
Sigma-Aldrich
Amyloid β Protein Fragment 1-42