Merck
  • Targeting ABL1 or ARG Tyrosine Kinases to Restrict HIV-1 Infection in Primary CD4+ T-Cells or in Humanized NSG Mice.
Targeting ABL1 or ARG Tyrosine Kinases to Restrict HIV-1 Infection in Primary CD4+ T-Cells or in Humanized NSG Mice.
Journal of acquired immune deficiency syndromes (1999) (2019-10-29)
Stephen D S McCarthy, Danila Leontyev, Pauline Nicoletti, Beth Binnington, Hannah N Kozlowski, Mario Ostrowski, Alan Cochrane, Donald R Branch, Raymond W Wong
ABSTRACT
Previous studies support dasatinib as a potent inhibitor of HIV-1 replication. However, a functional distinction between 2 kinase targets of the drug, ABL1 and ARG, has not been assessed. We used primary CD4 T-cells, CD8-depleted peripheral blood mononuclear cells (PBMCs) from a treatment naïve HIV-1 patient, and a humanized mouse model of HIV-1 infection. We assessed the roles of ABL1 and ARG during HIV-1 infection and use of dasatinib as a potential antiviral against HIV-1 in humanized mice. Primary CD4 T-cells were administered siRNA targeting ABL1 or ARG, then infected with HIV-1 containing luciferase reporter viruses. Quantitative polymerase chain reaction of viral integration of 4 HIV-1 strains was also assessed. CD8-depleted PBMCs were treated for 3 weeks with dasatinib. NSG mice were engrafted with CD34 pluripotent stem cells from human fetal cord blood, and infected with Ba-L virus after 19 weeks. Mice were treated daily with dasatinib starting 5 weeks after infection. siRNA knockdown of ABL1 or ARG had no effect on viral reverse transcripts, but increased 2-LTR circles 2- to 4-fold and reduced viral integration 2- to 12-fold. siRNA knockdown of ARG increased SAMHD1 activation, whereas knockdown of either kinase reduced RNA polymerase II activation. Treating CD8-depleted PBMCs from a treatment-naïve patient with 50 nM of dasatinib for 3 weeks reduced p24 levels by 99.8%. Ba-L (R5)-infected mice injected daily with dasatinib showed a 95.1% reduction in plasma viral load after 2 weeks of treatment. We demonstrate a novel nuclear role for ABL1 and ARG in ex vivo infection experiments, and proof-of-principle use of dasatinib in a humanized mouse model of HIV-1 infection.
Sigma-Aldrich
Dasatinib
Sigma-Aldrich
MISSION® esiRNA, targeting human ABL1