- On-line trypsin-encapsulated enzyme reactor by the sol-gel method integrated into capillary electrophoresis.
On-line trypsin-encapsulated enzyme reactor by the sol-gel method integrated into capillary electrophoresis.
A novel trypsin-encapsulation technique using the sol-gel method was developed for the preparation of an on-line enzyme reactor integrated into capillary electrophoresis. Trypsin was encapsulated in tetramethoxysilane-based hydrogel, and its enzymatic activity was evaluated using alpha-N-benzoyl-L-arginine ethyl ester and two peptides (bradykinin and [Ter8]-bradykinin). The enzyme encapsulation was carried out in a single step under mild conditions within a capillary, and 1.5-cm gel was formed at the inlet of the capillary. The resultant monolithic reactor showed excellent enzymatic activity, which was approximately 700 times higher than that in free solution, without stopping the flow. Separation of the unreacted substrates and products in the same capillary also showed high selectivity, and sample size in this system decreased 3 orders of magnitude from conventional tryptic reaction schemes. The encapsulated trypsin maintains its substrate specificity even in a sol-gel matrix. Furthermore, the encapsulated trypsin exhibits increased stability even after continuous use compared to that in free solution.