Merck
  • Extracellular transglutaminase 2 induces myotube hypertrophy through G protein-coupled receptor 56.
Extracellular transglutaminase 2 induces myotube hypertrophy through G protein-coupled receptor 56.
Biochimica et biophysica acta. Molecular cell research (2019-11-02)
Tomoya Kitakaze, Miki Yoshikawa, Yasuyuki Kobayashi, Naohiro Kimura, Naoki Goshima, Takahiro Ishikawa, Yoshiyuki Ogata, Yoko Yamashita, Hitoshi Ashida, Naoki Harada, Ryoichi Yamaji
ABSTRACT
Skeletal muscle secretes biologically active proteins that contribute to muscle hypertrophy in response to either exercise or dietary intake. The identification of skeletal muscle-secreted proteins that induces hypertrophy can provide critical information regarding skeletal muscle health. Dietary provitamin A, β-carotene, induces hypertrophy of the soleus muscle in mice. Here, we hypothesized that skeletal muscle produces hypertrophy-inducible secretory proteins via dietary β-carotene. Knockdown of retinoic acid receptor (RAR) γ inhibited the β-carotene-induced increase soleus muscle mass in mice. Using RNA sequencing, bioinformatic analyses, and literature searching, we predicted transglutaminase 2 (TG2) to be an all-trans retinoic acid (ATRA)-induced secretory protein in cultured C2C12 myotubes. Tg2 mRNA expression increased in ATRA- or β-carotene-stimulated myotubes and in the soleus muscle of β-carotene-treated mice. Knockdown of RARγ inhibited β-carotene-increased mRNA expression of Tg2 in the soleus muscle. ATRA increased endogenous TG2 levels in conditioned medium from myotubes. Extracellular TG2 promoted the phosphorylation of Akt, mechanistic target of rapamycin (mTOR), and ribosomal p70 S6 kinase (p70S6K), and inhibitors of mTOR, phosphatidylinositol 3-kinase, and Src (rapamycin, LY294002, and Src I1, respectively) inhibited TG2-increased phosphorylation of mTOR and p70S6K. Furthermore, extracellular TG2 promoted protein synthesis and hypertrophy in myotubes. TG2 mutant lacking transglutaminase activity exerted the same effects as wild-type TG2. Knockdown of G protein-coupled receptor 56 (GPR56) inhibited the effects of TG2 on mTOR signaling, protein synthesis, and hypertrophy. These results indicated that TG2 expression was upregulated through ATRA-mediated RARγ and that extracellular TG2 induced myotube hypertrophy by activating mTOR signaling-mediated protein synthesis through GPR56, independent of transglutaminase activity.
Sigma-Aldrich
MISSION® esiRNA, targeting human ADGRG1
Sigma-Aldrich
MISSION® esiRNA, targeting human LRP1