Merck
  • A randomized, double-blind, multicenter, phase 2b study to evaluate the safety and efficacy of a combination of tropifexor and cenicriviroc in patients with nonalcoholic steatohepatitis and liver fibrosis: Study design of the TANDEM trial.
A randomized, double-blind, multicenter, phase 2b study to evaluate the safety and efficacy of a combination of tropifexor and cenicriviroc in patients with nonalcoholic steatohepatitis and liver fibrosis: Study design of the TANDEM trial.
Contemporary clinical trials (2019-11-16)
Marcos Pedrosa, Star Seyedkazemi, Sven Francque, Arun Sanyal, Mary Rinella, Michael Charlton, Rohit Loomba, Vlad Ratziu, Jossy Kochuparampil, Laurent Fischer, Sujata Vaidyanathan, Quentin M Anstee
ABSTRACT
Nonalcoholic steatohepatitis (NASH) is a multifactorial disease involving different contributing mechanisms, with no approved therapies so far. Tropifexor (TXR), a farnesoid X receptor agonist, and cenicriviroc (CVC), a chemokine receptor types 2/5 antagonist, target the steatotic, inflammatory, and/or fibrotic pathways involved in NASH. TANDEM (CLJC242A2201J; NCT03517540) is a 48-week, phase 2b, randomized, double-blind, multicenter study in 200 adult patients with biopsy-proven NASH and liver fibrosis. Patients will be randomized in a 1:1:1:1 ratio to receive either TXR 140ā€ÆĪ¼g once daily (qd), CVC 150ā€Æmg qd, TXR 140ā€ÆĪ¼gā€Æ+ā€ÆCVC 150ā€Æmg qd, or TXR 90ā€ÆĪ¼gā€Æ+ā€ÆCVC 150ā€Æmg qd. The study comprises a 48-week treatment period and 4ā€Æweeks of follow-up. The key inclusion criterion is presence of NASH with fibrosis stage F2/F3 as seen on screening liver biopsy or on historical liver biopsy performed within 6ā€Æmonths prior to screening. The primary objective is evaluation of the safety and tolerability of combination therapy compared with the monotherapies over 48ā€Æweeks. The secondary objective is to evaluate efficacy as assessed by ā‰„1-point improvement in liver fibrosis versus baseline or resolution of steatohepatitis after 48ā€Æweeks. TANDEM will evaluate the combination of TXR and CVC with respect to safety and efficacy outcomes related to improvement in fibrosis or resolution of steatohepatitis. Given the effects of TXR and CVC in multiple pathophysiological pathways associated with NASH, combination therapy is likely to show additional benefits compared with monotherapy.
Product Description
Supelco
7Ī±-Hydroxy-4-cholesten-3-one, ≥95.0% (HPLC)