Merck
  • Grainyhead-like-2 confers NK-sensitivity through interactions with epigenetic modifiers.
Grainyhead-like-2 confers NK-sensitivity through interactions with epigenetic modifiers.
Molecular immunology (2018-12-06)
Ian MacFawn, Hannah Wilson, Luke A Selth, Ian Leighton, Ilya Serebriiskii, R Christopher Bleackley, Osama Elzamzamy, Joshua Farris, Phillip M Pifer, Jennifer Richer, Steven M Frisch
ABSTRACT
Natural Killer (NK) cells suppress tumor initiation and metastasis. Most carcinomas are heterogeneous mixtures of epithelial, mesenchymal and hybrid tumor cells, but the relationships of these phenotypes to NK susceptibility are understood incompletely. Grainyhead-like-2 (GRHL2) is a master programmer of the epithelial phenotype, that is obligatorily down-regulated during experimentally induced Epithelial-Mesenchymal Transition (EMT). Here, we utilize GRHL2 re-expression to discover unifying molecular mechanisms that link the epithelial phenotype with NK-sensitivity. GRHL2 enhanced the expression of ICAM-1, augmenting NK-target cell synaptogenesis and NK killing of target cells. The expression of multiple interferon response genes, including ICAM1, anti-correlated with EMT. We identified two novel GRHL2-interacting proteins, the histone methyltransferases KMT2C and KMT2D. Mesenchymal-epithelial transition, NK-sensitization and ICAM-1 expression were promoted by GRHL2-KMT2C/D interactions and by GRHL2 inhibition of p300, revealing novel and potentially targetable epigenetic mechanisms connecting the epithelial phenotype with target cell susceptibility to NK killing.
Sigma-Aldrich
Monoclonal Anti-GRHL2 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL3760, purified immunoglobulin, buffered aqueous glycerol solution