Merck
  • Impact of E2F-1 expression on clinical outcome of gastric adenocarcinoma patients with adjuvant chemoradiation therapy.
Impact of E2F-1 expression on clinical outcome of gastric adenocarcinoma patients with adjuvant chemoradiation therapy.
Clinical cancer research : an official journal of the American Association for Cancer Research (2008-01-04)
Jeeyun Lee, Cheol Keun Park, Joon Oh Park, Taekyu Lim, Young Suk Park, Ho Yeong Lim, Inkyoung Lee, Tae Sung Sohn, Jae Hyung Noh, Jin Seok Heo, Sung Kim, Do Hoon Lim, Kyoung-Mee Kim, Won Ki Kang
ABSTRACT
There are no reliable prognostic markers that identify gastric cancer patients who may benefit from adjuvant chemoradiation therapy. E2F-1 was shown to be associated with radiosensitivity and chemosensitivity in certain tumor types. Therefore, we analyzed expression and prognostic significance of E2F-1 along with thymidylate synthase (TS) in R(0)-resected gastric adenocarcinoma patients, who underwent adjuvant chemoradiation therapy with 5-fluorouracil (5-FU) and leucovorin. The chemosensitivity to 5-FU and radiosensitivity were tested in three E2F-1-overexpressed gastric cancer cell lines in vitro. The expressions of TS and E2F-1 were analyzed in 467 R(0)-resected primary gastric cancer patients, who received adjuvant chemoradiation therapy with 5-FU and leucovorin using tissue microarray. The E2F-1 immunopositivity rate was 22.2% (103 of 465 samples) with a cutoff value of 5% immunoreactivity, whereas the TS-positive expression occurred in 19.0% of the 463 tumors tested. Using stepwise Cox proportional hazards regression modeling, multivariate analyses showed that the E2F-1 immunopositivity predicted more favorable survival as compared with the E2F-1 immunonegativity with borderline statistical significance [P = 0.050, hazard ratio (HR) = 0.702, 95% confidence interval, 0.487, 1.013]. However, the E2F-1 immunopositivity did not retain its statistical significance at multivariate analysis for predicting disease-free survival (data not shown, P = 0.270), but stage was the only influential factor for disease-free survival in stages IB to IV (M(0)) patients (P < 0.001). TS immunopositivity did not influence survival (P = 0.459) or disease-free survival (P = 0.447). E2F-1 is a potentially novel independent prognostic factor that may identify gastric cancer patients who will likely benefit from adjuvant chemoradiation therapy following curative resection.
Sigma-Aldrich
Anti-Thymidylate Synthase Antibody, clone TS106, clone TS106, Chemicon®, from mouse
Pricing and availability is not currently available.