Merck
  • Radioprotective efficacy of tocopherol succinate is mediated through granulocyte-colony stimulating factor.
Radioprotective efficacy of tocopherol succinate is mediated through granulocyte-colony stimulating factor.
Cytokine (2011-09-02)
Pankaj K Singh, Stephen Y Wise, Elizabeth J Ducey, Darren S Brown, Vijay K Singh
ABSTRACT
The purpose of this study was to elucidate the role of granulocyte colony-stimulating factor (G-CSF) induced by α-tocopherol succinate (TS) in protecting mice from total-body irradiation. CD2F1 mice were injected with a radioprotective dose of TS and the levels of cytokine in serum induced by TS were determined by multiplex Luminex. Neutralization of G-CSF was accomplished by administration of a G-CSF antibody and confirmed by cytokine analysis. The role of G-CSF on gastrointestinal tissue protection afforded by TS after irradiation (11 Gy, 0.6 Gy/min of 60Co γ-radiation) was determined by analysis of jejunum histopathology for crypt, villi, mitotic figures, apoptosis, and cell proliferation. Our results demonstrate that TS protected mice against high doses of radiation-induced gastrointestinal damage and TS also induced very high levels of G-CSF and keratinocyte-derived chemokine (KC) production in peripheral blood 24 h after subcutaneous administration. When TS-injected mice were administered a neutralizing antibody to G-CSF, there was complete neutralization of G-CSF in circulating blood, and the protective effect of TS was significantly abrogated by G-CSF antibody. Histopathology of jejunum from TS-injected and irradiated mice demonstrated protection of gastrointestinal tissue, yet the protection was abrogated by administration of a G-CSF antibody. In conclusion, our current study suggests that induction of G-CSF resulting from TS administration is responsible for protection from 60Co γ-radiation injury.
Sigma-Aldrich
D-α-Tocopherol succinate, BioXtra, ≥98.0% (HPLC)
Sigma-Aldrich
D-α-Tocopherol succinate, semisynthetic, 1210 IU/g