Merck
  • CXCR4 regulates the early extravasation of metastatic tumor cells in vivo.
CXCR4 regulates the early extravasation of metastatic tumor cells in vivo.
Neoplasia (New York, N.Y.) (2009-07-02)
Peter Gassmann, Jörg Haier, Kerstin Schlüter, Britta Domikowsky, Claudia Wendel, Ulrike Wiesner, Robert Kubitza, Rainer Engers, Stephan W Schneider, Bernhard Homey, Anja Müller
ABSTRACT
Recent studies have demonstrated that the chemokine receptor CXCR4 plays a crucial role in organ-specific metastasis formation. Although a variety of studies showed the expression of chemokine receptors, in particular, CXCR4, by gastrointestinal tumors, the precise mechanisms of chemokine receptor-mediated homing of cancer cells to specific sites of metastasis remained elusive. Here, we used liver metastatic human HEP-G2 hepatoma and HT-29LMM colon cancer cells expressing functional CXCR4 to dissect the metastatic cascade by intravital fluorescence microscopy. Immunohistochemistry revealed that the CXCR4 ligand CXCL12 is expressed by endothelial cells and likely Kupffer cells lining the liver sinusoids. Tumor cell adhesion and extravasation in vivo was quantitatively analyzed using intravital fluorescence microscopy. Treatment of cells with an anti-CXCR4 antibody did not affect cell adhesion but significantly impaired tumor cell extravasation (HEP-G2; isotype control: 22.3% +/- 4.3% vs anti-CXCR4: 6.0% +/- 5.0%, P < .001). In addition, pretreatment of tumor cells with the ligand CXCL12 enhanced the activation of the small GTPases Rho, Rac, and cdc42 as well as tumor cell extravasation without affecting tumor cell adhesion within liver sinusoids. Taken together, the findings of the present study provide first in vivo insights into the early events of chemokine ligand/receptor-mediated liver metastasis formation of tumor cells and define tumor cell extravasation rather than tumor cell arrest as the rate-limiting event.
Sigma-Aldrich
Anti-Integrin β4 Antibody, clone ASC-3, clone ASC-3, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β4 Antibody, clone ASC-8, clone ASC-8, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin alpha-V Antibody, clone 272-17E6 (Azide free), clone 272-17E6, from mouse