Merck
  • Effect of intensified training on muscle ion kinetics, fatigue development, and repeated short-term performance in endurance-trained cyclists.
Effect of intensified training on muscle ion kinetics, fatigue development, and repeated short-term performance in endurance-trained cyclists.
American journal of physiology. Regulatory, integrative and comparative physiology (2013-07-26)
Thomas P Gunnarsson, Peter M Christensen, Martin Thomassen, Lars R Nielsen, Jens Bangsbo
ABSTRACT
The effects of intensified training in combination with a reduced training volume on muscle ion kinetics, transporters, and work capacity were examined. Eight well-trained cyclists replaced their regular training with speed-endurance training (12 × 30 s sprints) 2-3 times per week and aerobic high-intensity training (4-5 × 3-4 min at 90-100% of maximal heart rate) 1-2 times per week for 7 wk and reduced training volume by 70% (intervention period; IP). The duration of an intense exhaustive cycling bout (EX2; 368 ± 6 W), performed 2.5 min after a 2-min intense cycle bout (EX1), was longer (P < 0.05) after than before IP (4:16 ± 0:34 vs. 3:37 ± 0:28 min:s), and mean and peak power during a repeated sprint test improved (P < 0.05) by 4% and 3%, respectively. Femoral venous K(+) concentration in recovery from EX1 and EX2 was lowered (P < 0.05) after compared with before IP, whereas muscle interstitial K(+) concentration and net muscle K(+) release during exercise was unaltered. No changes in muscle lactate and H(+) release during and after EX1 and EX2 were observed, but the in vivo buffer capacity was higher (P < 0.05) after IP. Expression of the ATP-sensitive K(+) (KATP) channel (Kir6.2) decreased by IP, with no change in the strong inward rectifying K(+) channel (Kir2.1), muscle Na(+)-K(+) pump subunits, monocarboxylate transporters 1 and 4 (MCT1 and MCT4), and Na(+)/H(+) exchanger 1 (NHE1). In conclusion, 7 wk of intensified training with a reduced training volume improved performance during repeated intense exercise, which was associated with a greater muscle reuptake of K(+) and muscle buffer capacity but not with the amount of muscle ion transporters.
Sigma-Aldrich
Anti-Na+/K+ ATPase α-1 Antibody, clone C464.6, clone C464.6, Upstate®, from mouse
Sigma-Aldrich
Anti-Potassium Channel Kir2.1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Monocarboxylate Transporter 4 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Monocarboxylate Transporter 1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Na+K+ ATPase α-2 Antibody, serum, Upstate®