Merck
  • Fused Benzothiadiazole: A Building Block for n-Type Organic Acceptor to Achieve High-Performance Organic Solar Cells.
Fused Benzothiadiazole: A Building Block for n-Type Organic Acceptor to Achieve High-Performance Organic Solar Cells.
Advanced materials (Deerfield Beach, Fla.) (2019-03-19)
Jun Yuan, Yunqiang Zhang, Liuyang Zhou, Chujun Zhang, Tsz-Ki Lau, Guichuan Zhang, Xinhui Lu, Hin-Lap Yip, Shu Kong So, Serge Beaupré, Mathieu Mainville, Paul A Johnson, Mario Leclerc, Honggang Chen, Hongjian Peng, Yongfang Li, Yingping Zou
ABSTRACT
Narrow bandgap n-type organic semiconductors (n-OS) have attracted great attention in recent years as acceptors in organic solar cells (OSCs), due to their easily tuned absorption and electronic energy levels in comparison with fullerene acceptors. Herein, a new n-OS acceptor, Y5, with an electron-deficient-core-based fused structure is designed and synthesized, which exhibits a strong absorption in the 600-900 nm region with an extinction coefficient of 1.24 × 105 cm-1 , and an electron mobility of 2.11 × 10-4 cm2 V-1 s-1 . By blending Y5 with three types of common medium-bandgap polymers (J61, PBDB-T, and TTFQx-T1) as donors, all devices exhibit high short-circuit current densities over 20 mA cm-2 . As a result, the power conversion efficiency of the Y5-based OSCs with J61, TTFQx-T1, and PBDB-T reaches 11.0%, 13.1%, and 14.1%, respectively. This indicates that Y5 is a universal and highly efficient n-OS acceptor for applications in organic solar cells.
Sigma-Aldrich
Y5, ≥99%