Merck
  • Simultaneous High-Resolution Detection of Bioenergetic Molecules using Biomimetic-Receptor Nanopore.
Simultaneous High-Resolution Detection of Bioenergetic Molecules using Biomimetic-Receptor Nanopore.
Analytical chemistry (2019-10-31)
Zhuoqun Su, Yongfeng Wei, Xiao-Feng Kang
ABSTRACT
A novel artificial receptor, heptakis-[6-deoxy-6-(2-hydroxy-3-trimethylammonion-propyl) amino]-beta-cyclomaltoheptaose, with similar functions of mitochondrial ADP/ATP carrier protein, was synthesized and harbored in the engineered α-HL (M113R)7 nanopore, forming a single-molecule biosensor for sensing bioenergetic molecules and their transformations. The strategy significantly elevates both selectivity and signal-to-noise, which enables simultaneous recognition and detection of ATP, ADP, and AMP by real-time single-molecule measurement.
Product Description
Avanti
4ME 16:0 PC, Avanti Research - A Croda Brand
Avanti
4ME 16:0 PC, Avanti Research - A Croda Brand