Merck
  • New Bisoxazoline Ligands Enable Enantioselective Electrocatalytic Cyanofunctionalization of Vinylarenes.
New Bisoxazoline Ligands Enable Enantioselective Electrocatalytic Cyanofunctionalization of Vinylarenes.
Journal of the American Chemical Society (2019-09-10)
Niankai Fu, Lu Song, Jinjian Liu, Yifan Shen, Juno C Siu, Song Lin
ABSTRACT
In contrast to the rapid growth of synthetic electrochemistry in recent years, enantioselective catalytic methods powered by electricity remain rare. In this work, we report the development of a highly enantioselective method for the electrochemical cyanophosphinoylation of vinylarenes. A new family of serine-derived chiral bisoxazolines with ancillary coordination sites were identified as optimal ligands.
Sigma-Aldrich
sBOX(iPr), ≥95%
Sigma-Aldrich
sBOX(tBu), ≥95%