Merck
  • Anti-inflammatory effects of ABT-702, a novel non-nucleoside adenosine kinase inhibitor, in rat adjuvant arthritis.
Anti-inflammatory effects of ABT-702, a novel non-nucleoside adenosine kinase inhibitor, in rat adjuvant arthritis.
The Journal of pharmacology and experimental therapeutics (2001-02-13)
D L Boyle, E A Kowaluk, M F Jarvis, C H Lee, S S Bhagwat, M Williams, G S Firestein
ABSTRACT
Adenosine (ADO) is a homeostatic inhibitory autocoid that is released at sites of inflammation and tissue injury, and exerts anti-inflammatory effects via multiple interactions at ADO receptor subtypes. Inhibition of ADO kinase (AK) increases extracellular ADO concentrations and AK inhibitors have demonstrated ADO-mediated anti-inflammatory effects in acute models of inflammation. To evaluate the potential utility of this approach in chronic inflammation, a novel, potent, and selective non-nucleoside AK inhibitor, ABT-702, was tested in the rat adjuvant arthritis model. Animals were immunized with complete Freund's adjuvant on day 0 and were treated with vehicle or ABT-702 (20 mg/kg/b.i.d. p.o.) beginning on day 8. ABT-702 significantly inhibited arthritis as determined by paw volume. In addition, histologic and radiographic evidence of bone and cartilage destruction was significantly decreased in the treated group. Coadministration of the ADO receptor antagonist theophylline attenuated the anti-inflammatory effects of ABT-702, suggesting that this action was mediated through endogenous ADO release. To evaluate the mechanism of chondroprotection, Northern blot and electrophoretic mobility shift assays were performed on joints samples. These studies demonstrated that ABT-702 suppressed collagenase and stromelysin gene expression in treated animals. In addition, the activator protein-1 and nuclear factor-kappaB binding activity was also decreased. Therefore, ABT-702 inhibited clinical, radiographic, and histologic evidence of chronic inflammatory arthritis. The mechanism of joint protection is likely related to suppressed transcription factor activation and matrix metalloproteinase gene expression.
Product Description
Millipore
Fractogel® EMD SO3⁻ (S) Resin
Pricing and availability is not currently available.