Merck
  • Derivatives of Natural Product Agrimophol as Disruptors of Intrabacterial pH Homeostasis in Mycobacterium tuberculosis.
Derivatives of Natural Product Agrimophol as Disruptors of Intrabacterial pH Homeostasis in Mycobacterium tuberculosis.
ACS infectious diseases (2019-04-25)
Jie Wu, Ran Mu, Mingna Sun, Nan Zhao, Miaomiao Pan, Hongshuang Li, Yi Dong, Zhaogang Sun, Jie Bai, Minwan Hu, Carl F Nathan, Babak Javid, Gang Liu
ABSTRACT
This article reports the rational medicinal chemistry of a natural product, agrimophol (1), as a new disruptor of intrabacterial pH (pHIB) homeostasis in Mycobacterium tuberculosis (Mtb). Through the systematic investigation of the structure-activity relationship of 1, scaffold-hopping of the diphenylmethane scaffold, pharmacophore displacement strategies, and studies of the structure-metabolism relationship, a new derivative 5a was achieved. Compound 5a showed 100-fold increased potency in the ability to reduce pHIB to pH 6.0 and similarly improved mycobactericidal activity compared with 1 against both Mycobacterium bovis-BCG and Mtb. Compound 5a possessed improved metabolic stability in human liver microsomes and hepatocytes, lower cytotoxicity, higher selectivity index, and similar pKa value to natural 1. This study introduces a novel scaffold to an old drug, resulting in improved mycobactericidal activity through decreasing pHIB, and may contribute to the critical search for new agents to overcome drug resistance and persistence in the treatment of tuberculosis.