Merck
Evolution of Mechanisms that Control Mating in Drosophila Males.
Cell reports (2019-05-30)
Osama M Ahmed, Aram Avila-Herrera, Khin May Tun, Paula H Serpa, Justin Peng, Srinivas Parthasarathy, Jon-Michael Knapp, David L Stern, Graeme W Davis, Katherine S Pollard, Nirao M Shah
ABSTRACT
Genetically wired neural mechanisms inhibit mating between species because even naive animals rarely mate with other species. These mechanisms can evolve through changes in expression or function of key genes in sensory pathways or central circuits. Gr32a is a gustatory chemoreceptor that, in D. melanogaster, is essential to inhibit interspecies courtship and sense quinine. Similar to D. melanogaster, we find that D. simulans Gr32a is expressed in foreleg tarsi, sensorimotor appendages that inhibit interspecies courtship, and it is required to sense quinine. Nevertheless, Gr32a is not required to inhibit interspecies mating by D. simulans males. However, and similar to its function in D. melanogaster, Ppk25, a member of the Pickpocket family, promotes conspecific courtship in D. simulans. Together, we have identified distinct evolutionary mechanisms underlying chemosensory control of taste and courtship in closely related Drosophila species.