Merck
  • Surface analysis of sheep menisci after meniscectomy via infrared attenuated total reflection spectroscopy.
Surface analysis of sheep menisci after meniscectomy via infrared attenuated total reflection spectroscopy.
Journal of biophotonics (2019-03-20)
Ángela I López-Lorente, Pei Wang, Svenja Stein, Jonas Balko, Rui Lu, Lutz Dürselen, Boris Mizaikoff
ABSTRACT
Menisci are very important fibrocartilaginous tissue, which maintain biomechanical functions and physiological stabilization of knee joint. Meniscectomy is known as a surgery to recover partial functions from acute meniscus tears. However, the late consequences of total or partial meniscectomy include signs of osteoarthritis and even ligament instability. Infrared attenuated total reflection (IR-ATR) spectroscopy is a very useful technique, which can reveal molecular characteristics via the analysis of vibrational bands. The present study has employed IR-ATR spectroscopy to investigate sheep menisci samples after meniscectomy in a label-free fashion. Several differences of peak absorbance change and peak shift were observed between the native healthy samples and the meniscectomy samples in distinct IR wavenumber regions, such as amide I band, amide II band, C-H bending band as well as the sugar band region. Combining the results from the collagen protein IR spectra, it can be speculated that six months after meniscectomy collagen fibrils on the incision lose its ordered arrangement and a decrease in the triple helical structure of collagen fibril is observed. In addition, the collagen fibrils and proteoglycan content might also be slight varied after meniscectomy.
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture