Merck
  • TMCO1-mediated Ca2+ leak underlies osteoblast functions via CaMKII signaling.
TMCO1-mediated Ca2+ leak underlies osteoblast functions via CaMKII signaling.
Nature communications (2019-04-10)
Jianwei Li, Caizhi Liu, Yuheng Li, Qiaoxia Zheng, Youjia Xu, Beibei Liu, Weijia Sun, Yuan Li, Shuhui Ji, Mingwei Liu, Jing Zhang, Dingsheng Zhao, Ruikai Du, Zizhong Liu, Guohui Zhong, Cuiwei Sun, Yanqing Wang, Jinping Song, Shu Zhang, Jun Qin, Shukuan Ling, Xianhua Wang, Yingxian Li
ABSTRACT
Transmembrane and coiled-coil domains 1 (TMCO1) is a recently identified Ca2+ leak channel in the endoplasmic reticulum. TMCO1 dysfunction in humans is associated with dysmorphism, mental retardation, glaucoma and the occurrence of cancer. Here we show an essential role of TMCO1 in osteogenesis mediated by local Ca2+/CaMKII signaling in osteoblasts. TMCO1 levels were significantly decreased in bone from both osteoporosis patients and bone-loss mouse models. Tmco1-/- mice exhibited loss of bone mass and altered microarchitecture characteristic of osteoporosis. In the absence of TMCO1, decreased HDAC4 phosphorylation resulted in nuclear enrichment of HADC4, which leads to deacetylation and degradation of RUNX2, the master regulator of osteogenesis. We further demonstrate that TMCO1-mediated Ca2+ leak provides local Ca2+ signals to activate the CaMKII-HDAC4-RUNX2 signaling axis. The establishment of TMCO1 as a pivotal player in osteogenesis uncovers a novel potential therapeutic target for ameliorating osteoporosis.
Sigma-Aldrich
Anti-TMCO1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-WWP1 antibody produced in rabbit, affinity isolated antibody