- Hair pyrrole adducts serve as biomarkers for peripheral nerve impairment induced by 2,5-hexanedione and n-hexane in rats.
Hair pyrrole adducts serve as biomarkers for peripheral nerve impairment induced by 2,5-hexanedione and n-hexane in rats.
Pyrrole adducts are specific reaction products of 2,5-hexadione (2,5-HD) in vivo and are considered highly relevant to the pathogenesis of peripheral nerve impairments after exposure to n-hexane, though the exact mechanism remains unclear. In this study, 40 male Wistar rats were randomly divided into three experimental groups and one control group, in which all rat's hair were shaved completely at the beginning. The rats in three experimental groups were treated with 2,5-HD by gavage at dosages of 100, 200 and 300 mg/kg per day respectively, six times per week for 8 weeks. Abnormality of gait and changes in the rota-rod latency were surveilled. Pyrrole adducts in hair, urine and serum of all rats were measured at the endpoint. Results showed that the increased pyrrole adducts in hair, urine and serum accumulated in dose-response relationship. Spearman's correlation analysis between pyrrole adducts and gait scores showed that hair pyrrole adducts were highly relevant to the gait scores. Moreover, we treated rats with n-hexane and succeed to verify the results aforesaid. Further, multiply linear regression analysis showed that hair pyrrole adducts have higher partial correlation coefficients than these in serum and urine in both 2,5-HD and n-hexane treated models. Our findings draw the conclusion that the hair pyrrole adducts might serve as a promising biomarker of n-hexane induced peripheral neuropathy.