- Polyadenylic-polyuridylic acid plus locoregional radiotherapy versus chemotherapy with CMF in operable breast cancer: a 14 year follow-up analysis of a randomized trial of the Fédération Nationale des Centres de Lutte contre le Cancer (FNCLCC).
Polyadenylic-polyuridylic acid plus locoregional radiotherapy versus chemotherapy with CMF in operable breast cancer: a 14 year follow-up analysis of a randomized trial of the Fédération Nationale des Centres de Lutte contre le Cancer (FNCLCC).
Breast cancer research and treatment (2001-02-24)
A Laplanche, L Alzieu, T Delozier, J Berlie, C Veyret, P Fargeot, M Luboinski, J Lacour
PMID11194454
ABSTRACT
With a median follow-up of 14 years, the combination of polyadenylic-polyuridylic acid plus locoregional radiotherapy (257 patients) has significantly improved disease-free survival (p = 0.03) and significantly reduced the incidence of metastases (p = 0.04) when compared to CMF alone (260 patients), in women with operable breast cancer. The trial does not, however, permit an appreciation of the respective role of radiotherapy and PolyAU in these results.