Merck
  • Optimization of a nonradioactive method for consistent and sensitive determination of activated K-ras protein.
Optimization of a nonradioactive method for consistent and sensitive determination of activated K-ras protein.
Analytical biochemistry (2005-07-16)
Richard J Calvert, Wafa Kammouni, Keith D Kikawa
ABSTRACT
Accurate measurement of activity of wild-type K-ras protein is important due to its tumor suppressor action in tissues such as lung. A published method by Taylor and co-workers uses plasmid-containing Escherichia coli cells to produce a glutathione-S-transferase/raf-1 ras binding domain (GST-RBD) fusion protein attached to glutathione beads to isolate activated ras protein. We systematically optimized the method before use on lung tissues. Changing the GST-RBD protein induction temperature from the original 37 to 30 degrees C produced a consistently greater yield of fusion protein. To improve stability of the GST-RBD beads so as to perform large-scale experiments, 0.1% NaN(3) was added. NaN(3)-treated beads retained full affinity for at least 24 days. Sensitivity was improved by using a polyvinylidene difluoride membrane rather than nitrocellulose for immunoblotting. We also compared our GST-RBD beads with two commercial assay kits and found that our beads had both superior sensitivity and reduced variability. In summary, our modification of the GST-RBD affinity method to recover activated K-ras greatly increased the yield of fusion protein, prolonged the useful life of GST-RBD beads to at least 24 days, and enhanced detection sensitivity.