Merck
  • Sequential induction of 5-lipoxygenase gene expression and activity in Mono Mac 6 cells by transforming growth factor beta and 1,25-dihydroxyvitamin D3.
Sequential induction of 5-lipoxygenase gene expression and activity in Mono Mac 6 cells by transforming growth factor beta and 1,25-dihydroxyvitamin D3.
Proceedings of the National Academy of Sciences of the United States of America (1995-01-03)
M Brungs, O Rådmark, B Samuelsson, D Steinhilber
ABSTRACT
5-Lipoxygenase (5-LO; EC 1.13.11.34) activity in the human monocytic cell line Mono Mac 6 was upregulated by combined treatment with transforming growth factor beta 1 (TGF-beta) and 1,25-dihydroxyvitamin D3 (VD3). In undifferentiated cells, 5-LO enzyme activity was undetectable. After the addition of TGF-beta plus VD3, the activity of intact cells was 800 ng per 10(6) cells--500 times more than the assay detection limit. Also 5-LO protein and mRNA expression were induced > 128-fold and 64-fold, respectively, as compared to undifferentiated cells. Both TGF-beta and VD3 were required for these prominent responses. Either agent alone gave small amounts of 5-LO protein and mRNA but very low 5-LO activities. After the addition of TGF-beta and VD3, the induction of 5-LO protein was obvious after 1 day, but the increase in activity was delayed and did not appear until the second day. Pretreatment of cells with TGF-beta or VD3 alone for 2 days led to 5-LO protein expression but very low enzyme activity. Addition of the lacking second inducer was required for full induction of 5-LO protein expression and for upregulation of enzyme activity. Partial purification of 5-LO from Mono Mac 6 cells and recombination with soluble cellular proteins from different sources indicated the presence of cytosolic factors that affect the activity of 5-LO.