Merck
  • Life-threatening influenza pneumonitis in a child with inherited IRF9 deficiency.
Life-threatening influenza pneumonitis in a child with inherited IRF9 deficiency.
The Journal of experimental medicine (2018-08-26)
Nicholas Hernandez, Isabelle Melki, Huie Jing, Tanwir Habib, Susie S Y Huang, Jeffrey Danielson, Tomasz Kula, Scott Drutman, Serkan Belkaya, Vimel Rattina, Lazaro Lorenzo-Diaz, Anais Boulai, Yoann Rose, Naoki Kitabayashi, Mathieu P Rodero, Cecile Dumaine, Stéphane Blanche, Marie-Noëlle Lebras, Man Chun Leung, Lisa Sara Mathew, Bertrand Boisson, Shen-Ying Zhang, Stephanie Boisson-Dupuis, Silvia Giliani, Damien Chaussabel, Luigi D Notarangelo, Stephen J Elledge, Michael J Ciancanelli, Laurent Abel, Qian Zhang, Nico Marr, Yanick J Crow, Helen C Su, Jean-Laurent Casanova
ABSTRACT
Life-threatening pulmonary influenza can be caused by inborn errors of type I and III IFN immunity. We report a 5-yr-old child with severe pulmonary influenza at 2 yr. She is homozygous for a loss-of-function IRF9 allele. Her cells activate gamma-activated factor (GAF) STAT1 homodimers but not IFN-stimulated gene factor 3 (ISGF3) trimers (STAT1/STAT2/IRF9) in response to IFN-α2b. The transcriptome induced by IFN-α2b in the patient's cells is much narrower than that of control cells; however, induction of a subset of IFN-stimulated gene transcripts remains detectable. In vitro, the patient's cells do not control three respiratory viruses, influenza A virus (IAV), parainfluenza virus (PIV), and respiratory syncytial virus (RSV). These phenotypes are rescued by wild-type IRF9, whereas silencing IRF9 expression in control cells increases viral replication. However, the child has controlled various common viruses in vivo, including respiratory viruses other than IAV. Our findings show that human IRF9- and ISGF3-dependent type I and III IFN responsive pathways are essential for controlling IAV.
Sigma-Aldrich
MISSION® esiRNA, targeting human IRF9, RP11-468E2.4
Pricing and availability is not currently available.