Merck
  • Effects of a 15-month anti-TNF-α treatment on plasma levels of glycosaminoglycans in women with rheumatoid arthritis.
Effects of a 15-month anti-TNF-α treatment on plasma levels of glycosaminoglycans in women with rheumatoid arthritis.
Arthritis research & therapy (2018-09-20)
Anna Szeremeta, Agnieszka Jura-Półtorak, Ewa Maria Koźma, Andrzej Głowacki, Eugeniusz Józef Kucharz, Magdalena Kopeć-Mędrek, Krystyna Olczyk
ABSTRACT
In this study, the effect of 15-month anti-tumor necrosis factor alpha (TNF-α) treatment on circulating levels of plasma sulfated glycosaminoglycans (GAGs) and the nonsulfated GAG hyaluronic acid (HA) in female rheumatoid arthritis (RA) patients was assessed. Plasma was obtained from healthy subjects and RA women treated with TNF-α antagonists (etanercept or adalimumab or certolizumab pegol) in combination with methotrexate. GAGs were isolated from plasma samples using ion exchange low-pressure liquid chromatography. Total sulfated GAGs were quantified using a hexuronic acid assay. Plasma levels of keratan sulfate (KS) and HA were measured using immunoassay kits. Total sulfated GAGs and HA levels were higher in female RA patients before treatment in comparison to healthy subjects. KS levels did not differ between RA women and controls. Anti-TNF-α treatment resulted in normalization of plasma total GAG and HA levels in RA patients, without any effect on KS levels. Our results suggest that anti-TNF-α therapy has a beneficial effect on extracellular matrix remodeling in the course of RA.
Product Description
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
Protease from Streptomyces sp., Type XXI, ≥15 units/mg solid