Merck
  • Microtubules assemble near most kinetochores during early prometaphase in human cells.
Microtubules assemble near most kinetochores during early prometaphase in human cells.
The Journal of cell biology (2018-06-17)
Vitali Sikirzhytski, Fioranna Renda, Irina Tikhonenko, Valentin Magidson, Bruce F McEwen, Alexey Khodjakov
ABSTRACT
For proper segregation during cell division, each chromosome must connect to the poles of the spindle via microtubule bundles termed kinetochore fibers (K-fibers). K-fibers form by two distinct mechanisms: (1) capture of astral microtubules nucleated at the centrosome by the chromosomes' kinetochores or (2) attachment of kinetochores to noncentrosomal microtubules with subsequent transport of the minus ends of these microtubules toward the spindle poles. The relative contributions of these alternative mechanisms to normal spindle assembly remain unknown. In this study, we report that most kinetochores in human cells develop K-fibers via the second mechanism. Correlative light electron microscopy demonstrates that from the onset of spindle assembly, short randomly oriented noncentrosomal microtubules appear in the immediate vicinity of the kinetochores. Initially, these microtubules interact with the kinetochores laterally, but end-on attachments form rapidly in the first 3 min of prometaphase. Conversion from lateral to end-on interactions is impeded upon inhibition of the plus end-directed kinetochore-associated kinesin CenpE.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human CENPE
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Triton X-100, laboratory grade