Merck
  • Pharmacodynamic and pharmacokinetic characteristics of YMR-65, a tubulin inhibitor, in tumor-bearing mice.
Pharmacodynamic and pharmacokinetic characteristics of YMR-65, a tubulin inhibitor, in tumor-bearing mice.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2018-05-18)
Ali Fan, Jiali Wei, Mengru Yang, Qing Zhang, Yaliang Zhang, Qingwang Liu, Ning Li, Di Zhao, Yang Lu, Junxiu Li, Jie Zhao, Shuhua Deng, Bingjie Zhang, Hailiang Zhu, Xijing Chen
ABSTRACT
YMR-65​, 5-(5-bromo-1-methyl-1H-indol-3-yl)-3-(3-methoxyphenyl)-4, 5-dihydro-1H-pyrazole-1-carboxamide, is a potential tubulin inhibitor exhibiting good anticancer activity. In our study, we illustrated the biological activities in HepG2 cells and the pharmacodynamic and pharmacokinetic profiles were evaluated in murine H22 hepatoma-bearing mice. Molecular docking assay and colchicine competition assay indicated that YMR-65 could bind tightly to the colchicine binding site of tubulin. Further investigation demonstrated that YMR-65 arrested cells in the G2/M phase of cell cycle and induced apoptosis in HepG2 cells. Compared with control group, the tumor growth inhibition determined by final relative volume of tumor/the initial tumor volume were 32.57%, 24.00% and 34.95%, respectively, for YMR-65 (10 mg/kg), YMR-65 (20 mg/kg) and CA4P (10 m/kg) groups. Besides there were no obvious body change or tissue damage (enhanced by histopathology study). YMR-65 administration at 10 and 20 mg/kg in H22 tumor-bearing mice resulted in 1.87- and 1.80-fold longer half time (t1/2) and 0.36- and 0.78-fold lower area under concentration-time curve (AUC0-∞) in plasma in contrast with normal mice at 10 mg/kg. Furthermore, YMR-65 showed a wide distribution to various tissues or tumor and the highest distribution index (the ratio of AUCtissue or tumor/AUCplasma) was found in tumor, which implied that it might accumulate in tumor after administration. In brief, our results indicated that YMR-65 was a promising candidate with high antitumor efficacy and low tissue damage.
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride