Merck
  • Peroxidasin contributes to lung host defense by direct binding and killing of gram-negative bacteria.
Peroxidasin contributes to lung host defense by direct binding and killing of gram-negative bacteria.
PLoS pathogens (2018-05-19)
Ruizheng Shi, Zehong Cao, Hong Li, Jochen Graw, Guogang Zhang, Victor J Thannickal, Guangjie Cheng
ABSTRACT
Innate immune recognition is classically mediated by the interaction of host pattern-recognition receptors and pathogen-associated molecular patterns; this triggers a series of downstream signaling events that facilitate killing and elimination of invading pathogens. In this report, we provide the first evidence that peroxidasin (PXDN; also known as vascular peroxidase-1) directly binds to gram-negative bacteria and mediates bactericidal activity, thus, contributing to lung host defense. PXDN contains five leucine-rich repeats and four immunoglobulin domains, which allows for its interaction with lipopolysaccharide, a membrane component of gram-negative bacteria. Bactericidal activity of PXDN is mediated via its capacity to generate hypohalous acids. Deficiency of PXDN results in a failure to eradicate Pseudomonas aeruginosa and increased mortality in a murine model of Pseudomonas lung infection. These observations indicate that PXDN mediates previously unrecognized host defense functions against gram-negative bacterial pathogens.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Lipid A, diphosphoryl from Escherichia coli F583 (Rd mutant)