- Embryonic lethality and radiation hypersensitivity mediated by Rad51 in mice lacking Brca2.
Embryonic lethality and radiation hypersensitivity mediated by Rad51 in mice lacking Brca2.
Nature (1997-04-24)
S K Sharan, M Morimatsu, U Albrecht, D S Lim, E Regel, C Dinh, A Sands, G Eichele, P Hasty, A Bradley
PMID9126738
ABSTRACT
Inherited mutations in the human BRCA2 gene cause about half of the cases of early-onset breast cancer. The embryonic expression pattern of the mouse Brca2 gene is now defined and an interaction identified of the Brca2 protein with the DNA-repair protein Rad51. Developmental arrest in Brca2-deficient embryos, their radiation sensitivity, and the association of Brca2 with Rad51 indicate that Brca2 may be an essential cofactor in the Rad51-dependent DNA repair of double-strand breaks, thereby explaining the tumour-suppressor function of Brca2.