Merck
Control of lymphocyte function through CD27-CD70 interactions.
Seminars in immunology (1998-11-25)
S M Lens, K Tesselaar, M H van Oers, R A van Lier
ABSTRACT
In contrast to the expression of other TNFR/TNF family members, expression of CD27 and its ligand CD70 is predominantly confined to lymphocytes. High expression levels of CD27 appear to be dependent on proper ligation of antigen receptors, whereas for the induction of CD70 expression additional (co-stimulatory and/or pro-inflammatory) signals are required. Next to membrane-bound CD27 also a soluble form of CD27 is produced in the course of the immune response. Soluble CD27 (sCD27) is found in body fluids and can be used to monitor local and systemic immune activation. In addition, elevated serum concentrations of sCD27 are found in patients with B cell malignancies and levels of sCD27 strongly correlate with tumor load. Based on functional experiments and in vitro expression regulation data, we propose that interactions between activated lymphocytes in vivo can result in CD27-CD70 interactions that may regulate the size and function of antigen-primed lymphocyte populations.
Sigma-Aldrich
BMP-4 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
SCD27 LIGAND human, recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC)