Merck
  • Smart specimen preparation for freeze substitution and serial ultrathin sectioning of yeast cells.
Smart specimen preparation for freeze substitution and serial ultrathin sectioning of yeast cells.
Journal of electron microscopy (2009-03-18)
Masashi Yamaguchi, Hitoshi Okada, Yuichi Namiki
ABSTRACT
A smart and efficient method for freeze substitution and serial sectioning of yeast cells is described. Yeast cells were placed in a single layer between two copper disks, rapidly frozen, freeze substituted and embedded in an epoxy resin. The cell layer was re-embedded by the same resin, the surface trimmed leaving 1 mum above the cell layer, and serially sectioned. The sections were collected on the two-slit grids and placed on a Formvar film mounted to cover the holes of an aluminum supporting rack. The grids were removed from the rack, stained together using a silicon tube and observed in a transmission electron microscope. The images of yeast cells observed were clear and natural, and would be useful for a detailed 3D structural analysis such as structome.
Sigma-Aldrich
Epoxy embedding medium, accelerator, ≥95% (NT)
Pricing and availability is not currently available.