Merck
  • Soybean Hydrophobic Protein is Present in a Matrix Secreted by the Endocarp Epidermis during Seed Development.
Soybean Hydrophobic Protein is Present in a Matrix Secreted by the Endocarp Epidermis during Seed Development.
Scientific reports (2015-10-13)
Daryl E Enstone, Carol A Peterson, Mark Gijzen
ABSTRACT
Hydrophobic protein from soybean (HPS) is present in soybean dust and is an allergen (Gly m 1) that causes asthma in allergic individuals. Past studies have shown that HPS occurs on the seed surface. To determine the microscopic localization of HPS during seed development, monoclonal antibodies to HPS were used to visualize the protein by fluorescence and transmission electron microscopy. Seed coat and endocarp sections were also examined for pectin, cellulose, callose, starch, and protein by histochemical staining. HPS is present in the endocarp epidermal cells at 18 to 28 days post anthesis. At later stages of seed development, HPS occurs in extracellular secretions that accumulate unevenly on the endocarp epidermis and seed surface. HPS is synthesized by the endocarp epidermis and deposited on the seed surface as part of a heterogeneous matrix.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Gold antibody produced in goat, affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)
Pricing and availability is not currently available.