Merck
  • Clenbuterol residues in pig muscle after repeat administration in a growth-promoting dose.
Clenbuterol residues in pig muscle after repeat administration in a growth-promoting dose.
Meat science (2010-07-30)
Jelka Pleadin, Ana Vulić, Nina Persi, Nada Vahcić
ABSTRACT
The aim of this study was to determine the level of clenbuterol residues in muscle tissue of pigs after repeat administration in a growth-promoting dose. An anabolic dose of clenbuterol (20 μg/kg body mass per day) was administered orally to experimental group (n=12) for 28 days, whereas control animals (n=3) were left untreated. Clenbuterol treated pigs were randomly sacrificed (n=3) on days 0, 3, 7 and 14 of treatment discontinuation and clenbuterol residues determined in muscle tissue. Determination of residual clenbuterol was by enzyme-linked immunosorbent assay (ELISA) as a screening method and liquid chromatography tandem mass spectrometry (LC-MS/MS) as a confirmation method. The highest clenbuterol content in the muscle of treated animals was recorded on day 0 of treatment cessation (4.40±0.37 ng/g) and significantly (p<0.05) exceeded the maximum residue limit (MRL) of 0.1 ng/g. On day 3 of withdrawal, it was 0.49±0.22 ng/g and on day 7 0.10±0.02 ng/g (at MRL); on day 14 of treatment discontinuation, clenbuterol content was below the limit of detection (<0.1 ng/g) in all samples. Administration of clenbuterol as a growth promoter in pig production could lead to residues in meat for human consumption up to 7 days after treatment discontinuation.
Sigma-Aldrich
Clenbuterol hydrochloride, ≥95%