Zinc Acetate
Synonym(s):
Zinc acetate dihydrate
Linear Formula:
Zn(CH3COO)2 · 2H2O
CAS Number:
Molecular Weight:
219.51
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing