
Valpromide

Synonym(s):
2-Propylvaleramide, 2-propyl-pentanamide, Depamid, Depamide, Di-n-propylacetamide, Dipropylacetamide
Empirical Formula (Hill Notation):
C8H17NO
CAS Number:
Molecular Weight:
143.23
Products
Shipping
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing