Valaciclovir hydrochloride (anhydrous)
Synonym(s):
Valacyclovir hydrochloride
Empirical Formula (Hill Notation):
C13H20N6O4 · HCl
CAS Number:
Molecular Weight:
360.80
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing