Skip to Content
Merck
Urea-15N2 2 atom % 15N, 99% (CP)

Urea-15N2

Urea-15N2 2 atom % 15N, 99% (CP)
Synonym(s):
15N Labeled urea, Carbamide-15N2, Carbonyldiamide-15N2
Linear Formula:
H215NCO15NH2
CAS Number:
Molecular Weight:
62.04

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Application
Markush Class
Markush Group
Showing 1-5 of 5