Skip to Content
Merck
UK 14,304

UK 14,304

UK 14,304
Synonym(s):
5-Bromo-N-(2-imidazolin-2-yl)-6-quinoxalinamine, 5-Bromo-N-(4,5-dihydro-1H-imidazol-2-yl)-6-quinoxalinamine, Brimonidine
Empirical Formula (Hill Notation):
C11H10BrN5
CAS Number:
Molecular Weight:
292.13

Products

Product Category
Brand
Formula Weight
Gene Alias
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing