Skip to Content
Merck
Tris(4-carbazoyl-9-ylphenyl)amine ≥97%

Tris(4-carbazoyl-9-ylphenyl)amine

Tris(4-carbazoyl-9-ylphenyl)amine ≥97%
Synonym(s):
TCTA
Empirical Formula (Hill Notation):
C54H36N4
CAS Number:
Molecular Weight:
740.89

Products

Brand
Color
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing